KEY432442026 Key Organics / BIONET

(2RS,4RS)-4-(benzyloxy)-2-fluorocyclohexan-1-one relative stereochemistry

  • MDL number MFCD28155262
  • Cas Number : 1638612-61-9
  • Molecular Weight : 222.259

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/442219

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4870_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart