KEY444728204 Key Organics / BIONET

4-pentylcyclohexane-1-carboxylic acid

  • MDL number MFCD00466238
  • Cas Number : 38792-89-1
  • Molecular Weight : 198.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 56 - 60
Iupac Name 4-pentylcyclohexanecarboxylic acid
Inchi Code 1S/C12H22O2/c1-2-3-4-5-10-6-8-11(9-7-10)12(13)14/h10-11H,2-9H2,1H3,(H,13,14)
Inchi Key RVLAXPQGTRTHEV-UHFFFAOYSA-N

Description

Melting Point 56 - 60
Purity 90%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9049

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/12N-117_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart