KEY444728215 Key Organics / BIONET

1-benzyl-5-bromo-1,2-dihydropyridin-2-one

  • MDL number MFCD02326300
  • Cas Number : 217448-53-8
  • Molecular Weight : 264.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-benzyl-5-bromo-2(1H)-pyridinone
Inchi Code 1S/C12H10BrNO/c13-11-6-7-12(15)14(9-11)8-10-4-2-1-3-5-10/h1-7,9H,8H2
Inchi Key CTTIPOGAOKXCDF-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/453134

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/AS-3117_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart