KEY444728231 Key Organics / BIONET

7-chloro-6-methyl-2-(2,2,2-trifluoroacetyl)-1H,2H,3H,4H,5H,10H-benzo[b]1,6-naphthyridin-10-one

  • MDL number MFCD19705677
  • Cas Number : 1325307-44-5
  • Molecular Weight : 344.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/453150

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3661_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart