KEY444728237 Key Organics / BIONET

2-cyano-N-[4-(trifluoromethyl)phenyl]acetamide

  • MDL number MFCD05669245
  • Cas Number : 24522-30-3
  • Molecular Weight : 228.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-cyano-N-[4-(trifluoromethyl)phenyl]acetamide
Inchi Code 1S/C10H7F3N2O/c11-10(12,13)7-1-3-8(4-2-7)15-9(16)5-6-14/h1-4H,5H2,(H,15,16)
Inchi Key JBNCFFDGYDZEEN-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/453156

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3667_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart