KEY444728267 Key Organics / BIONET

1-(3,5-dimethylphenyl)-4-oxo-1,4-dihydropyridazine-3-carboxylic acid

  • MDL number MFCD08753943
  • Cas Number : 937670-77-4
  • Molecular Weight : 244.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/453186

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3698_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart