KEY444728272 Key Organics / BIONET

7-(4-bromophenyl)-3H,4H-thieno[3,2-d]pyrimidin-4-one

  • MDL number MFCD04973921
  • Cas Number : 827614-31-3
  • Molecular Weight : 307.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/453191

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3703_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart