KEY444728276 Key Organics / BIONET

7-(3-methylphenyl)-3H,4H-thieno[3,2-d]pyrimidin-4-one

  • MDL number MFCD16639687
  • Cas Number : 1031989-23-7
  • Molecular Weight : 242.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

signalword Warning
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart