KEY444728283 Key Organics / BIONET

3-(2,4-dimethylbenzenesulfonyl)-6,7-difluoro-1,4-dihydroquinolin-4-one

  • MDL number MFCD19705866
  • Cas Number : 1325304-15-1
  • Molecular Weight : 349.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/453202

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3714_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart