KEY444728285 Key Organics / BIONET

3-(3-chlorobenzenesulfonyl)-1-ethyl-6,7-difluoro-1,4-dihydroquinolin-4-one

  • MDL number MFCD20753679
  • Cas Number : 1326863-28-8
  • Molecular Weight : 383.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/453204

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3716_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart