KEY444728469 Key Organics / BIONET

4-chloro-6-(pyrrolidin-1-yl)-2-(trifluoromethyl)pyrimidine

  • MDL number MFCD09743688
  • Cas Number : 1189872-15-8
  • Molecular Weight : 251.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at -18°C
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/443877

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P302 + P352,P304 + P340,P305 + P351 + P338,P402 + P404,P411
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/DS-0200_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart