KEY444728608 Key Organics / BIONET

2-methyl-2H,4H,5H,6H,7H-pyrazolo[4,3-c]pyridine dihydrochloride

  • MDL number MFCD09999162
  • Cas Number : 157327-45-2
  • Molecular Weight : 210.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-methyl-4,5,6,7-tetrahydro-2H-pyrazolo[4,3-c]pyridine dihydrochloride
Inchi Code 1S/C7H11N3.2ClH/c1-10-5-6-4-8-3-2-7(6)9-10;;/h5,8H,2-4H2,1H3;2*1H
Inchi Key ABBAYPXCQSWDQG-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Store at 4
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/444016

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P302 + P352,P304 + P340,P305 + P351 + P338,P402 + P404,P411
MSDS https://www.keyorganics.net/media/catalog/product/sds/DS-0355_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart