KEY444728883 Key Organics / BIONET

tert-butyl N-(5-bromo-1,3,4-thiadiazol-2-yl)carbamate

  • MDL number MFCD11848382
  • Cas Number : 1048358-33-3
  • Molecular Weight : 280.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 5-bromo-1,3,4-thiadiazol-2-ylcarbamate
Inchi Code 1S/C7H10BrN3O2S/c1-7(2,3)13-6(12)9-5-11-10-4(8)14-5/h1-3H3,(H,9,11,12)
Inchi Key FVENMKPZBGIYPA-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/444291

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/DS-0657_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart