KEY444728979 Key Organics / BIONET

2,4-dichloro-7-(4-methylbenzenesulfonyl)-7H-pyrrolo[2,3-d]pyrimidine

  • MDL number MFCD13193624
  • Cas Number : 934524-10-4
  • Molecular Weight : 342.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2,4-dichloro-7-[(4-methylphenyl)sulfonyl]-7H-pyrrolo[2,3-d]pyrimidine
Inchi Code 1S/C13H9Cl2N3O2S/c1-8-2-4-9(5-3-8)21(19,20)18-7-6-10-11(14)16-13(15)17-12(10)18/h2-7H,1H3
Inchi Key DTDGVNQSPAWHTH-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/444387

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/DS-0760_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart