KEY444728993 Key Organics / BIONET

2-chloro-5-iodo-7H-pyrrolo[2,3-d]pyrimidine

  • MDL number MFCD13189363
  • Cas Number : 1060815-90-8
  • Molecular Weight : 279.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-chloro-5-iodo-7H-pyrrolo[2,3-d]pyrimidine
Inchi Code 1S/C6H3ClIN3/c7-6-10-1-3-4(8)2-9-5(3)11-6/h1-2H,(H,9,10,11)
Inchi Key CPGBAXNIILKLJF-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/444401

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P302 + P352,P304 + P340,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/DS-0775_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart