KEY444729399 Key Organics / BIONET

methyl 2-(bromomethyl)-3-chlorobenzoate

  • MDL number MFCD04114322
  • Cas Number : 188187-03-3
  • Molecular Weight : 263.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/444807

Safety Information

signalword Danger
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H314,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/DS-1195_SDS.pdf
pictograms GHS05,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart