KEY444729521 Key Organics / BIONET

tert-butyl 3-oxo-1-oxa-8-azaspiro[4.5]decane-8-carboxylate

  • MDL number MFCD10001421
  • Cas Number : 954236-44-3
  • Molecular Weight : 255.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 3-oxo-1-oxa-8-azaspiro[4.5]decane-8-carboxylate
Inchi Code 1S/C13H21NO4/c1-12(2,3)18-11(16)14-6-4-13(5-7-14)8-10(15)9-17-13/h4-9H2,1-3H3
Inchi Key XDSCHYPLQWGQRC-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/444929
Availability details for
Quantity
  • Availability
  • Add to Cart