KEY444729563 Key Organics / BIONET

benzyl 4-(2-methoxy-2-oxoethyl)piperidine-1-carboxylate

  • MDL number MFCD02179006
  • Cas Number : 170737-53-8
  • Molecular Weight : 291.347

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/444971

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/DS-1380_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart