KEY444729661 Key Organics / BIONET

3-methyl-5-phenyl-1,2-oxazole-4-carboxylic acid

  • MDL number MFCD03086120
  • Cas Number : 17153-21-8
  • Molecular Weight : 203.197

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store under N2
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/445069

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404,P422
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/DS-1486_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart