KEY444730050 Key Organics / BIONET

1-(cyclopentylmethyl)piperazine

  • MDL number MFCD08677888
  • Cas Number : 82500-22-9
  • Molecular Weight : 168.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(cyclopentylmethyl)piperazine
Inchi Code 1S/C10H20N2/c1-2-4-10(3-1)9-12-7-5-11-6-8-12/h10-11H,1-9H2
Inchi Key HDSJFNAPZZCQAT-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Liquid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/445458

Safety Information

signalword Danger
pcodes P260,P262,P270,P280,P302 + P352,P304 + P340,P305 + P351 + P338,P402 + P404,P403 + P235,P411
MSDS https://www.keyorganics.net/media/catalog/product/sds/DS-1898_SDS.pdf
hcodes H226,H302,H312,H314,H315,H319,H331,H332
pictograms GHS05,GHS06,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart