KEY444730059 Key Organics / BIONET

2,4-dichloro-6-methoxyquinoline

  • MDL number MFCD03285967
  • Cas Number : 70049-46-6
  • Molecular Weight : 228.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store under N2 at 4
Physical Form White to Dark Brown Solid
Shipping Temperature Store under N2 at 4
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/445467

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P302 + P352,P304 + P340,P305 + P351 + P338,P402 + P404,P411,P422
MSDS https://www.keyorganics.net/media/catalog/product/sds/DS-1908_SDS.pdf
pictograms GHS07
hstatements H302,H312,H315,H319,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart