KEY444730544 Key Organics / BIONET

5-nitro-1H-indole-3-carbonitrile

  • MDL number MFCD02090219
  • Cas Number : 7147-14-0
  • Molecular Weight : 187.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-nitro-1H-indole-3-carbonitrile
Inchi Code 1S/C9H5N3O2/c10-4-6-5-11-9-2-1-7(12(13)14)3-8(6)9/h1-3,5,11H
Inchi Key AKLPDARNWZLNPV-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/445952

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P302 + P352,P304 + P340,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/DS-2430_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart