KEY444730727 Key Organics / BIONET

30,34-dihydroxynonacyclo[18.10.2.2^{2,5}.0^{3,16}.0^{4,13}.0^{6,11}.0^{17,31}.0^{22,27}.0^{28,32}]tetratriaconta-1(30),2,4,6(11),7,9,13,15,17,19,22,24,26,28,31,33-hexadecaene-12,21-dione

  • MDL number MFCD00034580
  • Cas Number : 128-59-6
  • Molecular Weight : 488.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.80
Storage temp. RT
Physical Form Grey to Black Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/446135

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/media/catalog/product/sds/DS-2619_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart