KEY444730731 Key Organics / BIONET

(2-methylpyridin-3-yl)methanamine

  • MDL number MFCD03411326
  • Cas Number : 58539-64-3
  • Molecular Weight : 122.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2-methyl-3-pyridinyl)methanamine
Inchi Code 1S/C7H10N2/c1-6-7(5-8)3-2-4-9-6/h2-4H,5,8H2,1H3
Inchi Key YMROUWDKBBTAPS-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/446139

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/DS-2623_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart