KEY444731089 Key Organics / BIONET

4-methyl-2-(trifluoromethyl)pyrimidine

  • MDL number MFCD09881360
  • Cas Number : 1017464-05-9
  • Molecular Weight : 162.115

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/446497

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/DS-3005_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart