KEY444731111 Key Organics / BIONET

3-chloro-2-(chloromethyl)pyridine hydrochloride

  • MDL number MFCD20527153
  • Cas Number : 124425-87-2
  • Molecular Weight : 198.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-chloro-2-(chloromethyl)pyridine hydrochloride
Inchi Code 1S/C6H5Cl2N.ClH/c7-4-6-5(8)2-1-3-9-6;/h1-3H,4H2;1H
Inchi Key WOWKPCRREBTZCL-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/446519

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/DS-3028_SDS.pdf
hcodes H302,H312,H314,H315,H319,H332,H335
pictograms GHS05,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart