KEY444731124 Key Organics / BIONET

methyl[1-(2-phenyladamantan-1-yl)propan-2-yl]amine hydrochloride

  • MDL number MFCD01674041
  • Cas Number : 31897-80-0
  • Molecular Weight : 319.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form White to Off-white Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/446532

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P302 + P352,P304 + P340,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/media/catalog/product/sds/DS-3041_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart