KEY444731127 Key Organics / BIONET

methyl 2-amino-2-cyclopropylacetate hydrochloride

  • MDL number MFCD12031219
  • Cas Number : 535936-86-8
  • Molecular Weight : 165.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl amino(cyclopropyl)acetate hydrochloride
Inchi Code 1S/C6H11NO2.ClH/c1-9-6(8)5(7)4-2-3-4;/h4-5H,2-3,7H2,1H3;1H
Inchi Key SRMSADQSMWKRRW-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/446535

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P302 + P352,P304 + P340,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/DS-3044_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart