KEY444731187 Key Organics / BIONET

tert-butyl 3-(2-ethoxy-2-oxoethyl)azetidine-1-carboxylate

  • MDL number MFCD11616049
  • Cas Number : 158602-35-8
  • Molecular Weight : 243.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 3-(2-ethoxy-2-oxoethyl)-1-azetidinecarboxylate
Inchi Code 1S/C12H21NO4/c1-5-16-10(14)6-9-7-13(8-9)11(15)17-12(2,3)4/h9H,5-8H2,1-4H3
Inchi Key HTYZZXUWPRCEIN-UHFFFAOYSA-N

Description

Purity 97%
Physical Form Liquid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/446595

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/DS-3106_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart