KEY444731323 Key Organics / BIONET

phenyl pyrrolidine-1-carboxylate

  • MDL number MFCD00276291
  • Cas Number : 55379-71-0
  • Molecular Weight : 191.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/453219

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/FA-0016_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart