KEY444731343 Key Organics / BIONET

N-(6-chloropyridazin-3-yl)acetamide

  • MDL number MFCD09835313
  • Cas Number : 14959-31-0
  • Molecular Weight : 171.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-(6-chloro-3-pyridazinyl)acetamide
Inchi Code 1S/C6H6ClN3O/c1-4(11)8-6-3-2-5(7)9-10-6/h2-3H,1H3,(H,8,10,11)
Inchi Key UNQTYWWIVXWOJT-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/453239

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/SS-4877_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart