KEY444731344 Key Organics / BIONET

5-(5-bromofuran-2-yl)-3-(4-fluorophenyl)-1,2,4-oxadiazole

  • MDL number MFCD08068598
  • Cas Number : 915896-25-2
  • Molecular Weight : 309.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/453240

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/SS-4878_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart