KEY444731349 Key Organics / BIONET

(oxan-4-ylidene)amino 4-methylbenzene-1-sulfonate

  • MDL number MFCD28347979
  • Cas Number : 61108-59-6
  • Molecular Weight : 269.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store under N2
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/453245

Safety Information

signalword Warning
pcodes P422, P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/SS-4883_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart