KEY444790312 Key Organics / BIONET

2-[6-(benzyloxy)naphthalen-2-yl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

  • MDL number MFCD22493794
  • Cas Number : 1313367-62-2
  • Molecular Weight : 360.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(6-(benzyloxy)naphthalen-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Inchi Code 1S/C23H25BO3/c1-22(2)23(3,4)27-24(26-22)20-12-10-19-15-21(13-11-18(19)14-20)25-16-17-8-6-5-7-9-17/h5-15H,16H2,1-4H3
Inchi Key UVJOREZPMMEFQU-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin GB
Availability details for
Quantity
  • Availability
  • Add to Cart