KEY444790330 Key Organics / BIONET

ethyl 5-fluoropyridine-2-carboxylate

  • MDL number MFCD11616895
  • Cas Number : 148541-70-2
  • Molecular Weight : 169.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 5-fluoro-2-pyridinecarboxylate
Inchi Code 1S/C8H8FNO2/c1-2-12-8(11)7-4-3-6(9)5-10-7/h3-5H,2H2,1H3
Inchi Key AHMGOYHQXHQSQE-UHFFFAOYSA-N

Description

Purity 97%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/454368

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/KK-0202_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart