KEY444790351 Key Organics / BIONET

[(2S)-4-benzylmorpholin-2-yl]methanamine

  • MDL number MFCD18632720
  • Cas Number : 186293-55-0
  • Molecular Weight : 206.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name [(2S)-4-benzyl-2-morpholinyl]methanamine
Inchi Code 1S/C12H18N2O/c13-8-12-10-14(6-7-15-12)9-11-4-2-1-3-5-11/h1-5,12H,6-10,13H2/t12-/m0/s1
Inchi Key CKZVBXBEDDAEFE-LBPRGKRZSA-N

Description

Purity 95%
Storage temp. Store under N2 at 4
Physical Form Liquid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/454389

Safety Information

signalword Warning
pcodes P411,P422, P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/SS-4885_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart