KEY444790363 Key Organics / BIONET

tert-butyl 6-oxo-2-azaspiro[3.3]heptane-2-carboxylate

  • MDL number MFCD15071430
  • Cas Number : 1181816-12-5
  • Molecular Weight : 211.261

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/454401

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4897_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart