KEY448420334 Key Organics / BIONET

1-phenyl-4-{[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl}piperazine

  • MDL number MFCD16294543
  • Cas Number : 2096339-39-6
  • Molecular Weight : 378.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link http://www.keyorganics.net/bionet/catalog/product/view/id/488163

Safety Information

signalword Warning
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart