KEY448420554 Key Organics / BIONET

2-chloro-3-nitro-5-(trifluoromethyl)pyridine

  • MDL number MFCD06255171
  • Cas Number : 72587-15-6
  • Molecular Weight : 226.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-chloro-3-nitro-5-(trifluoromethyl)pyridine
Inchi Code 1S/C6H2ClF3N2O2/c7-5-4(12(13)14)1-3(2-11-5)6(8,9)10/h1-2H
Inchi Key DRPIKFKCAJGTJF-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Liquid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/488575

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/CS-10667_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart