KEY448894158 Key Organics / BIONET

2-chlorobenzene-1-carboximidamide hydrochloride

  • MDL number MFCD00275607
  • Cas Number : 18637-02-0
  • Molecular Weight : 191.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/512916

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2J-038_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart