KEY448894174 Key Organics / BIONET

4-{2-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]ethyl}morpholine

  • MDL number MFCD04440855
  • Cas Number : 364794-81-0
  • Molecular Weight : 317.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-{2-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]ethyl}morpholine
Inchi Code 1S/C18H28BNO3/c1-17(2)18(3,4)23-19(22-17)16-7-5-15(6-8-16)9-10-20-11-13-21-14-12-20/h5-8H,9-14H2,1-4H3
Inchi Key OOGUDBLEQZQAGC-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/512932

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/AS-7078_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart