KEY448894175 Key Organics / BIONET

4-chloro-3-iodo-1-[(4-methoxyphenyl)methyl]-1H-pyrazolo[4,3-c]pyridine

  • MDL number MFCD19689569
  • Cas Number : 1246349-97-2
  • Molecular Weight : 399.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-chloro-3-iodo-1-(4-methoxybenzyl)-1H-pyrazolo[4,3-c]pyridine
Inchi Code 1S/C14H11ClIN3O/c1-20-10-4-2-9(3-5-10)8-19-11-6-7-17-13(15)12(11)14(16)18-19/h2-7H,8H2,1H3
Inchi Key LTPHGFLBWHINDY-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/512933

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/BF-0054_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart