KEY448894180 Key Organics / BIONET

5-nitro-2,3-dihydro-1-benzofuran

  • MDL number MFCD22376562
  • Cas Number : 17403-47-3
  • Molecular Weight : 165.148

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/512936

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/CL-0049_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart