KEY448894254 Key Organics / BIONET

2-hydroxy-N-methylbenzamide

  • MDL number MFCD00045757
  • Cas Number : 1862-88-0
  • Molecular Weight : 151.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-hydroxy-N-methylbenzamide
Inchi Code 1S/C8H9NO2/c1-9-8(11)6-4-2-3-5-7(6)10/h2-5,10H,1H3,(H,9,11)
Inchi Key BCKXMQIYWWTZDP-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/512950

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/HF-0760_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart