KEY448894268 Key Organics / BIONET

4-methyl-5-(3-methylphenyl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD02228635
  • Cas Number : 333313-78-3
  • Molecular Weight : 205.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-methyl-5-(3-methylphenyl)-2,4-dihydro-3H-1,2,4-triazole-3-thione
Inchi Code 1S/C10H11N3S/c1-7-4-3-5-8(6-7)9-11-12-10(14)13(9)2/h3-6H,1-2H3,(H,12,14)
Inchi Key AGSMLYNSNAEUPT-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/100537

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/JS-2645_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart