KEY448894282 Key Organics / BIONET

tert-butyl 2-oxo-6-azaspiro[3.4]octane-6-carboxylate

  • MDL number MFCD17016202
  • Cas Number : 203661-71-6
  • Molecular Weight : 225.288

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/512974

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4905_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart