KEY454861345 Key Organics / BIONET

(9H-fluoren-9-yl)methyl N-[(2R)-4-methyl-1-oxopentan-2-yl]carbamate

  • MDL number MFCD01318544
  • Cas Number : 146803-42-1
  • Molecular Weight : 337.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 71 - 73
Purity 0.90
Storage temp. RT
Physical Form Off-white to Dark Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/441530

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/NH-0706_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart