KEY454861417 Key Organics / BIONET

3,5-dimethyl-1H-pyrazol-4-amine

  • MDL number MFCD00187666
  • Cas Number : 5272-86-6
  • Molecular Weight : 111.148

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 203
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/516933

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/DL-0069_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart