KEY454861493 Key Organics / BIONET

1-(benzenesulfonyl)-1H-indole-3-carbaldehyde

  • MDL number MFCD02681985
  • Cas Number : 80360-20-9
  • Molecular Weight : 285.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 157-161 C
Iupac Name 1-(phenylsulfonyl)-1H-indole-3-carbaldehyde
Inchi Code 1S/C15H11NO3S/c17-11-12-10-16(15-9-5-4-8-14(12)15)20(18,19)13-6-2-1-3-7-13/h1-11H
Inchi Key ZLARITBCJILRBL-UHFFFAOYSA-N

Description

Melting Point 157-161 C
Purity 97%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/516986

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/NS-00010_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart