KEY456787251 Key Organics / BIONET

(3,4-dimethylphenyl)boronic acid

  • MDL number MFCD01009694
  • Cas Number : 55499-43-9
  • Molecular Weight : 149.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store under N2
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/519646

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P302 + P352,P304 + P340,P305 + P351 + P338,P402 + P404,P422
hcodes H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/media/catalog/product/sds/AS-3161_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart